Learn More
General Laboratory Products National Food Laboratory (NFL) 8WM-7166-100
Supplier:  General Laboratory Products 8WM7166100
Encompass_Preferred
National Food Laboratory (NFL) Aseptic Validation Medium - 100ml, 8 oz. wide mouth glass bottle, 12 per case. For the selective and differential isolation of Pseudomonas sp. using CN or CFC supplementation.
Catalog No. NC2402228