Learn More
Kuhner Shaker Inc STERILE SINGLE-USE BAG
Supplier:  Kuhner Shaker Inc NSMX710004
Encompass
One Standard Sterile Single-Use Bag for use with Kuhner Shaker's SB200-X Orbitally Shaken Bioreactors. Includes bio-weldable tubing and integrated (non-invasive) pH and DO Optical Sensors. (H)
Catalog No. 50-257-0326