Learn More
Hanna Pharmaceutical Supply Co., Inc. STERILE WATER INJ 10ML
Supplier: Hanna Pharmaceutical Supply Co., Inc. 0409488710
10 ml Fliptop Vial Sterile, nonpyrogenic, solute-free preparation of distilled water for injection. For use only as a sterile solvent or diluent vehicle for drugs or solutions suitable for parenteral administration pH 5.5 (5.0-7.0). Contains no bacteriostat, antimicrobial agent or added buffer 25/PK
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.