Learn More
MilliporeSigma STERITEST VENTS
Supplier:  MilliporeSigma TEFG02525
Encompass
Steritest Vent, 25micrometer Dia., 0.2micrometer membrane, PTFE, needle attached, use for venting glass vials with rubber septum tops and rigid plastic vials also for sterility testing of ampoules or collapsible bags. 25/PK.
Catalog No. NC9409756