Learn More
Cayman Chemical StevIolbIosIde 10mg
SDP
Supplier:  Cayman Chemical 3651310 MG
Encompass_Preferred
A natural non-caloric sweetener and an active metabolite of rebaudioside A; inhibits the proliferation of MDA-MB-231 cells at 250 UG/ml; active against M. tuberculosis (MIC = 3.75 UG/ml)
Catalog No. 50-291-1841