Learn More
Medchemexpress LLC Stevioside | 57817-89-7 | 804.87 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN066950MG
Encompass_Preferred
Stevioside is an orally active sweetener that can be isolated from Stevia rebaudiana. It exhibits several beneficial activities, including antihypertensive, antihyperglycemic, antioxidant, anti-inflammatory, and anti-tumor properties.
  • Antihypertensive activity
  • Antihyperglycemic activity
  • Antioxidant activity
  • Anti-inflammatory activity
  • Anti-tumor activities
Catalog No. 50-003-88463