Learn More
Medchemexpress LLC Stevioside (Standard) | 57817-89-7 | 804.87 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0669R
Encompass_Preferred
Stevioside (Standard) is the analytical standard of Stevioside, intended for research and analytical applications. It is an orally active sweetener isolated from Stevia rebaudiana, possessing antihypertensive, antihyperglycemic, antioxidant, anti-inflammatory, and anti-tumor activities.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Acts as an orally active sweetener.
  • Exhibits antihypertensive properties.
  • Shows antihyperglycemic activity.
  • Functions as an antioxidant.
  • Possesses anti-inflammatory effects.
  • Demonstrates anti-tumor activities.
Catalog No. 50-003-90235