Learn More
Medchemexpress LLC HY-15845 10mg , STF-083010 CAS:307543-71-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1584510MG
Encompass_Preferred
HY-15845 10mg STF-083010 CAS: 307543-71-1 STF-083010 is a specific IRE1α inhibitor. STF-083010 inhibits Ire1 endonuclease activity, without affecting its kinase activity, after endoplasmic reticulum stress. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0954