Learn More
Abcam STF-083010, IRE1alpha endonuclease inhibitor, 25MG
Supplier: Abcam AB14617625MG
MW 317.4 Da. Selective IRE1α endonuclease inhibitor. Dose dependently inhibits XBP-1 splicing. Shows cancer cell selective cytotoxic and cytostatic antiproliferative effects. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement