Learn More
Medchemexpress LLC STF-31 | 724741-75-7 | 99.79% | 423.53 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY18728
Encompass_Preferred
STF-31 is a selective inhibitor of glucose transporter 1 (GLUT1) with an IC50 of 1 μM. It also functions as a NAMPT inhibitor, inhibiting glucose uptake in renal cell carcinoma (RCC) 4 cells.
  • Selective inhibitor of glucose transporter 1 (GLUT1)
  • Functions as a NAMPT inhibitor
  • Inhibits glucose uptake in renal cell carcinoma (RCC) 4 cells
Catalog No. 50-003-85382