Learn More
Med Vet International Stifle Distractor w/ Spinlock, X-Small, J0240WBS
Supplier:  Med Vet International JOR240WBS
Encompass_Preferred
The working tips on our spinlock are the same as the ratchet version. The spinlock offers more secure distraction as it is less likely to slip. The spinlock also offers an infinite range of position versus the ratchet.
Catalog No. 50-005-85318