Learn More
TARGETMOL CHEMICALS INC STING agonist-1 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T832825MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. STING agonist-1 (G10) is a human-specific agent that elicits antiviral activity against emerging Alphaviruses.
Catalog No. 50-287-6765