Learn More
Medchemexpress LLC STING agonist-12 | 2259624-71-8 | MFCD34676739 | 98.3% | 490.88 g/mol | C25H19ClF4N2O2 | 5 MG

Supplier: Medchemexpress LLC HY1470105MG
STING agonist-12 is a non-nucleotide, orally active small-molecule activator of human STING intended for research use. It demonstrates potent STING activation in cell-based assays and is provided as a high-purity solid suitable for in vitro and in vivo studies.
- Activates human STING (EC50 185 nM).
- Orally active small molecule suitable for in vivo studies.
- High purity: 98.3%.
- Molecular weight 490.88 g/mol; formula C25H19ClF4N2O2.
- Soluble in DMSO at 250 mg/mL; ultrasonic assistance recommended.
- Store protected from light; short-term 4°C; in solvent: -80°C for 6 months, -20°C for 1 month.
- Supplied in mg-scale quantities for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.