Learn More
Medchemexpress LLC Sting agonist-16 | 652142-94-4 | ≥99.0% | 396.23 | C19H11Cl2N5O | 5 MG

Supplier: Medchemexpress LLC HY1319945MG
STING agonist-16 is a small-molecule activator of the STING (stimulator of interferon genes) pathway used as a research tool to study innate immune signaling, antiviral responses, and antitumor immunity. It is supplied as a high-purity solid suitable for biochemical and cellular assays.
- Activates STING signaling in vitro.
- High purity ≥99.0%.
- Molecular formula C19H11Cl2N5O; molecular weight 396.23.
- Off-white to light brown solid appearance.
- Powder storage: -20°C for long-term stability.
- Available in small research-sized quantities for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.