Learn More
Medchemexpress LLC HY-103665 10mg Medchemexpress, STING agonist-3 CAS:2138299-29-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10366510MG
Encompass_Preferred
Medchemexpress, HY-103665 10mg STING agonist-3 CAS:2138299-29-1 STING agonist-3, extracted from patent WO2017175147A1 (example 10), is a selective and non-nucleotide small-molecule STING agonist with a pEC50 and pIC50 of 7.5 and 9.5, respectively. STING agonist-3 has durable anti-tumor effect and tremendous potential to improve treatment of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8155