Learn More
Medchemexpress LLC STING agonist-4 | 2138300-40-8 | 95.1% | 678.74 g/mol | C34H38N12O4 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1239435MG
Encompass_Preferred
STING agonist-4 is a small-molecule agonist of the Stimulator of Interferon Genes (STING) receptor intended for research use. It activates STING signaling and is used in cell-based assays to study innate immune responses and inflammation.
  • Potent STING receptor agonist with apparent IC50 of 20 nM.
  • Binds full-length STING with an apparent Kd of ~1.6 nM.
  • Induces dose-dependent IFN-β secretion (EC50 = 3.1 μM) in cell assays.
  • Molecular formula C34H38N12O4 and molecular weight 678.74 g/mol.
  • White to gray solid; soluble in DMSO (~1 mg/mL), poorly soluble in water.
  • Powder stored at -20°C under inert gas; solutions stored at -80°C for long term.
Catalog No. 50-002-35163