Learn More
Genetex Inc STING Monoclonal Antibody [MSVA-515M] HistoMAX™ (GTX04484) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04484
Encompass_Preferred
The STING antibody [MSVA-515M] HistoMAX™ (GTX04484) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human mucosa from appendix tissue using GTX04484 STING antibody [MSVA-515M] HistoMAX. STING staining of inflammatory cells predominates in the interfollicular zone STING immunohistochemistry.
Catalog No. NC3667796