Learn More
Medchemexpress LLC HY-138682 10mg , STING-IN-2 CAS:346691-38-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13868210MG
Encompass_Preferred
HY-138682 10mg STING-IN-2 CAS: 346691-38-1 STING-IN-2 (C-170) is a potent and covalent STING inhibitor. STING-IN-2 efficiently inhibits both mouse STING (mmSTING) and human STING (hsSTING). STING-IN-2 can be used for autoinflammatory disease research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9510