Learn More
Selleck Chemical LLC STING inhibitor C-178
SDP
Supplier:  Selleck Chemical LLC S666725MG
Encompass_Preferred
C-178 is a covalent inhibitor of STING covalently bind to Cys91
Catalog No. 50-000-02359