Learn More
Chemglass Life Sciences Chemglass Magnetic PTFE Stir Bar, Turbo Rare-Earth, Elliptical, 15 x 10 mm
SDP
Supplier:  Chemglass Life Sciences CG2005T02
Encompass_Preferred
Chemglass magnetic PTFE stir bar with a rare-earth turbo magnet and an elliptical (egg-shaped) profile. It provides exceptional magnetic coupling with near-total resistance to demagnetization and is identified by an inert carbon black spot. Made in the United Kingdom.
  • PTFE construction.
  • Rare-earth turbo magnet for exceptional magnetic strength.
  • Dimensions: 15 x 10 mm.
  • Elliptical (egg-shaped) profile.
  • Identified by inert carbon black spot.
  • Exceptional resistance to demagnetization.
  • Made in the United Kingdom.
Catalog No. NC1810973