Learn More
Chemglass Life Sciences Stir Bar, Magnetic, Triangular, PTFE, 20mm L X 8mm Diameter
SDP
Supplier:  Chemglass Life Sciences CG200911
Encompass_Preferred
Stir Bar, Magnetic, Triangular, PTFE, 20mm L X 8mm Diameter. Effective for dissolving solids and mixing sediments in a flat bottom container such as a beaker or Erlenmeyer flask. Manufactured with FDA-grade and USP Class VI PTFE.
Catalog No. 50-872-475