Learn More
Ace Glass, Inc. Stirbar, PTFE, v-vane, fits 3mL and 5mL V-vials with 14/10joints
SDP
Supplier:  Ace Glass, Inc. 1366803
Encompass
Stirbar, PTFE, v-vane, fits 3mL and 5mL V-vials with 14/10joints
Catalog No. NC1056864