Learn More
Chemglass Life Sciences Chemglass Stir Bar Retriever, PTFE, 150 x 10 mm - for retrieving magnetic stir bars
SDP
Supplier:  Chemglass Life Sciences CG2015T01
Encompass_Preferred
The Chemglass stir bar retriever is a PTFE pickup rod designed to safely remove magnetic stir bars from laboratory vessels. Molded from pure, FDA-approved PTFE, it provides chemical inertness and is autoclavable for repeated use.
  • Dimensions: 150 x 10 mm.
  • Material: pure, FDA-approved PTFE.
  • Autoclavable for sterilization.
  • Chemically inert and leak-free construction.
  • Designed to retrieve magnetic stir bars without contamination.
Catalog No. NC2583358