Learn More
Analytical Sales & Services Stir Bars 1.6X4.8MM, Parylene-Encapsulated Stainless Steel, Non-Magnetic, 1000 Units
Supplier:  Analytical Sales & Services 13352
Encompass
STIR BARS 1.6X4.8MM 1000CS stir bars are engineered for precise agitation in photoredox and parallel synthesis systems. Made with durable parylene-encapsulated stainless steel, they are chemical resistant and non-magnetic, ensuring controlled mixing for sensitive applications. Packaged with 1000 units, these stir bars support high-throughput experiments, distinguishing themselves from others with their efficient manipulation by external magnetic forces.
Catalog No. NC3760901