Learn More
Analytical Sales & Services Stir Bars, Parylene Encapsulated Stainless Steel, 96- and 24-Well Compatibility, 1000 units
Supplier:  Analytical Sales & Services 13258
Encompass
Introducing Stir Bars, designed for photoredox and parallel synthesis reaction block systems, which enhance mixing in high-throughput experimentation (HTE). These stir bars are made of parylene encapsulated stainless steel, allowing magnetic stirring functionality while remaining non-magnetic themselves. The unique cylinder shape and precise dimensions enable compatibility with 96- and 24-well configurations, making them an ideal choice for efficient chemical reactions and experiments.
Catalog No. NC1671248