Learn More
Wilmad Labglass STIR BLADE PTFE 1/8IN 19X48MM
Supplier:  Wilmad Labglass LG9510T100
Encompass
Stirrer Blade, PTFE. For Shaft OD: 10mm; Blade Length x Width: 19mm x 48mm; Fits Flask Capacity: 100mL; Minimum [ST] Joint Clearance: 24/40. Made of 1/8 in. polytetrafluoroethylene (PTFE). For 10mm OD Stirrer shafts with side buttons.
Catalog No. 50-807-952