Learn More
Wilmad Labglass STIR BLADE PTFE 1/8IN 19X75MM
Supplier:  Wilmad Labglass LG9510T104
Encompass
Stirrer Blade, PTFE. For Shaft OD: 10mm; Blade Length x Width: 19mm x 75mm; Fits Flask Capacity: 500mL to 1000mL; Minimum [ST] Joint Clearance: 24/40. Made of 1/8 in. polytetrafluoroethylene (PTFE)
Catalog No. 50-803-304