Learn More
Wilmad Labglass STIR BLADE PTFE 1/8IN 24X130MM
Supplier:  Wilmad Labglass LG9510T108
Encompass
Stirrer Blade, PTFE. For Shaft OD: 10mm; Blade Length x Width: 24mm x 1308mm; Fits Flask Capacity: 5000mL; Minimum [ST] Joint Clearance: 29/42. Made of 1/8 in. polytetrafluoroethylene (PTFE). For 10mm OD Stirrer shafts with side butto
Catalog No. 50-807-770