Learn More
Ace Glass, Inc. Stir blade, stainless, 23x113mm, 10mm shaft
SDP
Supplier:  Ace Glass, Inc. 808608
Encompass_Preferred
23X113mm 304 SS stir blades for 10mm O.D. button type stir shafts. For compatible stir shafts see ACE 8068, 8070, 8074, and 8075 series.
Catalog No. NC0751861