Learn More
Wilmad Labglass STIR CHSPKE BRNG ONLY 45/50
Supplier:  Wilmad Labglass LG9470206
Encompass_Preferred
Stirrer Bearing, Chesapeake, 10mm. Joint Size: [ST] 45/50. Grease-free PTFE Stirrer Bearing designed for moderate vacuum applications or for use at atmospheric pressure.
Catalog No. 50-806-037