Learn More
V&P Scientific Inc STIR ELEMENT, DISC, NdFeB Magnet, 48MGO, Parylene C Coated, Used in Syringes, Tubes/Vials, 10/pack
Supplier:  V&P Scientific Inc VP 782N6
Encompass
STIR ELEMENT, DISC, NdFeB Magnet, 48MGO, Parylene C Coated, 6.4mm Diameter, 0.7mm Height, 80°C Max Operating Temp for Magnets
Catalog No. NC0003616