Learn More
Wilmad Labglass STIR SHAFT GLAS HD 50L
Supplier:  Wilmad Labglass LG9504110
Encompass_Preferred
Stirrer Shaft, Button Style, Glass Rod. Shaft O.D.: 19mm; Overall Length: 820mm; Fits Flask Capacity: 50000mL. Precision ground with side button for attaching LG-9515-series or LG-9516T-series Stirrer blades.
Catalog No. 50-807-279