Learn More
V&P Scientific Inc STIR ELEMENT, SANDWICH STYLE TUNING FORK - WITH PIVOT POINT WINGS, NdFeB Magnet, 48MGO, PTFE Plate, Used in Microplates, Tubes/Vials, 10/pack
Supplier:  V&P Scientific Inc VP 772FN616TFP
Encompass
STIR ELEMENT, SANDWICH STYLE TUNING FORK - WITH PIVOT POINT WINGS, NdFeB Magnet, 48MGO, PTFE Plate, 16.0mm Length, 6.1mm Width, 1.6mm Height, 80°C Max Operating Temp for Magnets
Catalog No. NC2212072