Learn More
Ace Glass, Inc. Stirbar, PTFE, egg shape, 50.8mmx19.1mm
SDP
Supplier:  Ace Glass, Inc. 1366313
Encompass
50.8mm x 19.1mm egg-shaped PTFE magnetic stir bar which provides a pivot point without a separate ring or abrupt change in contour. Designed to be used with round bottom flasks and DynaBlocs.
Catalog No. 50-983-533