Learn More
Chemglass Life Sciences Stirrer Blade, PTFE, Banana Shape, Perforated, 10mm, for 500mL Flasks, 34/45 minimum joint clearance
SDP
Supplier:  Chemglass Life Sciences CG208110
Encompass_Preferred
Stirrer Blade, PTFE, Banana Shape, Perforated, 10mm, for 500mL Flasks, 34/45 minimum joint clearance. Blade is shaped to resemble the contour of a round bottom flask. Blades are perforated. Made of 1/8" PTFE to fit 10mm OD stirrer shafts. Minimum joint size listed is the size required to allow passage of the blade.
Catalog No. CG208110