Learn More
Chemglass Life Sciences Stirrer Blade, PTFE, 10mm, for 100mL to 500mL Flasks, 19mm Width x 48mm Length, 24/40 minimum joint clearance
SDP
Supplier:  Chemglass Life Sciences CG208001
Encompass_Preferred
Stirrer Blade, PTFE, 10mm, for 100mL to 500mL Flasks, 19mm Width x 48mm Length, 24/40 minimum joint clearance. Stirrer blade made of 1/8" thick solid PTFE for use with both Chem-Stir™ and button style stirring shafts having a 10mm OD.
Catalog No. CG208001