Learn More
Chemglass Life Sciences Stirrer, Magnetic, Two Position, Dura-Mag, 120V
SDP
Supplier:  Chemglass Life Sciences CLS410002
Encompass
Dura-Mag digital, front-end, magnetic stirrers are designed for both cell culture in spinner flasks as well as general laboratory procedures requiring the use of magnetic stirring equipment.Dura-Mag stirrers feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation.Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance.Dura-Mag 2-position, low profile chassis measure 9.5" W x 22.5" D x 3.5” H(front to back measurement includes power cord and control knob) and are designed to fit side-by-side in most incubators.Maximum flask size is (2) 8L vessels.Optional factory installed motion alarm available.Speed range: 25-400RPM.Please note: An Optional Flask Centering Template for Corning Disposable 500mL Spinner flasks is available.Installing the drop on template ensures the Corning Disposable Spinner flasks are centered directly over the magnet for optimal stirring.
Catalog No. 50-143-399