Learn More
Electron Microscopy Sciences Stirring Rod Solid PTFE Rod, 200 mm, 8 mm diameter
SDP
Supplier:  Electron Microscopy Sciences 6620120
Encompass_Preferred
PTFE solid rod with tapered end, rods are inert and will not scratch. Use up to 280 deg C..
OAL: 200 mm
Rod: 8 mm
Catalog No. 50-326-40