Learn More
Chemglass Life Sciences Stirring Shaft, PTFE Covered SS, 510mm OAL, 81mm OD Anchor Style Blade, Fits 1000mL Reactors
SDP
Supplier:  Chemglass Life Sciences CG2078X100
Encompass_Preferred
Stirring Shaft, PTFE Covered SS, 510mm OAL, 81mm OD Anchor Style Blade, Fits 1000mL Reactors
Catalog No. NC0831218