Learn More
Sino Biological Recombinant Mouse WEE2 Protein (Full Length, GST Tag), Active
Supplier:  Sino Biological W0610G500
Encompass_Preferred
Recombinant full length mouse WEE2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-2556