Learn More
MSC PIP Cut Resistant Gloves, 18-Gauge, Nitrile Foam Coated, Size 7, 1/Pr.
Supplier:  MSC 37092343
Encompass
These lightweight, 18-gauge, fiberglass-free gloves provide excellent tactile sensitivity and cut resistance. The seamless knit liner is constructed from recycled polyester, steel fiber, and spandex for comfort, while the foam nitrile coating on the palm and fingers disperses fluids for an improved grip. Featuring ESD-safe construction and touchscreen compatibility, these gloves are suitable for diverse industrial environments including assembly, maintenance, and logistics.
  • Breathable and durable foam nitrile coating for superior grip
  • Touchscreen compatible design allows device operation without removal
  • Reinforced thumb crotch for enhanced durability
  • ESD-safe construction suitable for static-sensitive applications
  • Complies with FDA food handling requirements 21 CFR, Part 177
  • Seamless 18-gauge liner provides comfort and dexterity
  • Silicone-free material composition
Catalog No. 50-005-08803