Learn More
MSC Value Collection Steel Square, 10-1/2 in. Blade, 6-3/8 in. Base, Beveled Edge, 1 ea.
Supplier: MSC 00540427
This steel square is a high-precision dimensional measuring tool designed for accurate layout and inspection tasks. Constructed from durable steel, it features a 10-1/2 in. blade and a 6-3/8 in. base, providing stability and reliability for professional applications.
- Accuracy of 0.0004 in.
- Beveled edge design for improved visibility and precision
- Robust steel construction