Learn More
Selleck Chemical LLC STM2457 5mg
SDP
Supplier:  Selleck Chemical LLC S98705MG
Encompass_Preferred
STM2457 is a highly potent and selective first-in-class catalytic inhibitor of RNA Methyltransferase METTL3 with an IC50 of 16.9 nM. STM2457 is highly specific for METTL3 and showed no inhibition of other RNA methyltransferases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3514