Learn More
Selleck Chemical LLC STM2457 S9870-100MG
SDP
Supplier:  Selleck Chemical LLC S9870100MG
Encompass_Preferred
STM2457 is a highly potent and selective first-in-class catalytic inhibitor of RNA Methyltransferase METTL3 with an IC50 of 16 9 nM STM2457 is highly specific for METTL3 and showed no inhibition of other RNA methyltransferases
Catalog No. 50-000-24700