Learn More
Med Vet International Pitch-It IV Stand, Floor Stand, 2-Hook, Three Leg, 30007-012, 12/Case
Supplier:  Med Vet International 881335CS
Encompass_Preferred
When no longer needed, it can be discarded into recycling Tripod design for stability Made of sturdy aluminum material Lightwieght for easy portablility Not for use with IV bags exceeding 1200 mL or with a pump
Catalog No. 50-304-6848