Learn More
Cayman Chemical STng AgonIst 12b 1mg
SDP
Supplier:  Cayman Chemical 325211 MG
Encompass_Preferred
A STING agonist; binds to STING (Kd = 26.4 µM); induces interferon reporter gene expression in cells expressing human or mouse STING (EC50s = 7.45 and 10.23 µM, respectively); induces expression of inflammatory cytokines in THP-1 cells at 40 µM
Catalog No. 50-294-7610