Learn More
Cayman Chemical STng M284 varInthumnre 100ug
SDP
Supplier:  Cayman Chemical 23594100 UG
Encompass_Preferred
Source: N-terminal His-tagged human recombinant protein purified from E. coli • MW: 28.8 kDa
Catalog No. 50-299-5328