Learn More
Cayman Chemical STng R232 varInthumnre 100ug
SDP
Supplier:  Cayman Chemical 22816100 UG
Encompass_Preferred
Source: N-terminal His-tagged STING catalytic domain purified from E. coli • MW: ~28.8 kDa
Catalog No. 50-299-4040