Learn More
Enzo Life Sciences STO-609 (5mg). CAS: 1173022-21-3
SDP
Supplier:  Enzo Life Sciences BMLEI3890005
Encompass_Preferred
STO-609 is a specific CaM kinase kinase (CaM-KK) inhibitor. Ki=0.2μM and 0.04μM for α and β isotypes respectively. Demonstrates >80-fold selectivity for CAM-KK over CaMK1, CaMK2, CaMK4, MLCK, PKA, PKC, and p42 MAPK. Concentrations up to 10μg/ml do not affect cellular viability. It is a tool for exploring the role of the CaM-KK pathway in various physiological processes. Alternative name: 7-Oxo-7H-benzimidazo[2,1-a]benz[de]isoquinoline-3-carboxylic acid, acetate salt. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0765