Learn More
Selleck Chemical LLC STO-609 5mg 52029-86-4
SDP
Supplier:  Selleck Chemical LLC S82745MG
Encompass_Preferred
STO-609 is a specific inhibitor of the Ca2+/Calmodulin-dependent protein kinase kinase(CaM-KK) that inhibits the activities of recombinant CaM-KK? and CaM-KK? isoforms, with Ki values of 80 and 15 ng/ml, respectively, and also inhibits their autophosphorylation activities. STO-609 inhibits AMPKK activity and inhibits autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5877